Nilfisk Straight Steel Wand for GM80, 23" L, 32mm dia

Representative image only.

Nilfisk Straight Steel Wand for GM80, 23" L, 32mm dia

Nilfisk | Mfr: 11112401| Item: 8656467

Enhance the performance of your vacuum

₹12,900.00INR / Each

₹12,900.00INR / Each
Product Description

Key Features

  • 2 year warranty

More About this Item

Keep your vacuum running at its best with these replacement parts and accessories.

Two year warranty
Specifications
  • Compatible WithGM80
  • Length (InchShort)23
  • Diameter (cm)3.2
  • DescriptionStraight Steel Wand for GM80, 23" L, 32mm dia
  • Warranty2 year
Reviews
Q&A