Nilfisk Flexible Rubber Crevice Nozzle for GM80, 32mm

Representative image only.

Nilfisk Flexible Rubber Crevice Nozzle for GM80, 32mm

Nilfisk | Mfr: 81140900| Item: 8656464

Enhance the performance of your vacuum

₹5,100.00INR / Each

₹5,100.00INR / Each
Product Description

More About this Item

Keep your vacuum running at its best with these replacement parts and accessories.

Specifications
  • Compatible WithGM80
  • Diameter (cm)3.2
  • DescriptionFlexible Rubber Crevice Nozzle for GM80, 32mm
Reviews
Q&A