Nilfisk 1408658500 Replacement Main Filter for Eliminator II
Representative image only.
Nilfisk 1408658500 Replacement Main Filter for Eliminator II
Nilfisk | Mfr: 140 8658 500| Item: 8656453
Collect a large volume of dry material directly into the tank
₹37,900.00INR / Each
₹37,900.00INR / Each
Product Description
Key Features
  • Use only for collecting dry materials
  • Serves as a pre-filter for the HEPA filter to extend the life of the HEPA filter
More About this Item
Use the filter when using the dust bag to collect dry materials. The float basket and or HEPA filter must be installed behind the main filter to prevent the main filter from collapsing during use.
Specifications
  • Compatible WithEliminator II vacuum
  • DescriptionReplacement main Filter for Eliminator II