National Marker HW5W Safety Labels, Non-Hazardous Waste, 6" x 6", Adhesive Vinyl; 25/Pk

Representative image only.

National Marker HW5W Safety Labels, Non-Hazardous Waste, 6" x 6", Adhesive Vinyl; 25/Pk

National Marker | Mfr: HW5W| Item: 8646310

Identify hazardous waste to comply with EPA and DOT regulations

₹8,400.00INR / Pkg of 25

₹8,400.00INR / Pkg of 25
Product Description

Key Features

  • 4-mil thick vinyl labels offer excellent durability
  • Easy-to-use self-adhesive labels
  • Green label indicates non-hazardous waste
  • Measures 6" x 6"

More About this Item

Appropriately identify hazardous waste to comply with EPA and DOT regulations. Self-adhesive labels are made of 4 mil, pressure-sensitive vinyl for better durability. Write in the information needed for your particular use. Compliance:,EPA 40CFR, DOT 49CFR

Specifications
  • HeaderNON-HAZARDOUS
  • LanguagesEnglish
  • MaterialPaper
  • Width (in)6
  • Height (in)6
  • DescriptionSafety Labels, Non-Hazardous Waste, 6" x 6", Adhesive Vinyl; 25/Pk
Reviews
Q&A