National Marker HM27 Hazardous Materials Identification Label, 4" X 4"

Representative image only.

National Marker HM27 Hazardous Materials Identification Label, 4" X 4"

National Marker | Mfr: HM27| Item: 8639601

Provide immediate indication of chemical hazards

₹7,700.00INR / Roll of 250

₹7,700.00INR / Roll of 250
Product Description

Key Features

  • Self-adhesive labels
  • Easy to write on surface
  • OSHA 29 CFR1910.1200 compliant

More About this Item

Right-to-know hazardous communication labels provide an immediate indication of chemical hazards in containers. Labels are self-adhesive and made of pressure sensitive paper. Information can easily be written on surface.

Specifications
  • HeaderHAZARDOUS
  • LanguagesEnglish
  • MaterialPaper
  • ComplianceOSHA 29 CFR1910.1200
  • Width (in)4
  • Height (in)4
  • DescriptionHazardous Materials Identification Label, 4" X 4"
Reviews
Q&A