National Marker DL4AL D.O.T. Shipping Labels, Flammable Liquid, 4" x 4"; 500/Roll

Representative image only.

National Marker DL4AL D.O.T. Shipping Labels, Flammable Liquid, 4" x 4"; 500/Roll

National Marker | Mfr: DL4AL| Item: 8639702

₹12,100.00INR / Roll of 500

₹12,100.00INR / Roll of 500
Product Description

More About this Item

Clearly identify hazardous materials during shipment in compliance with DOT regulations. All labels are made of pressure sensitive paper for easy application. Shipping labels measure 4" x 4" and are available in a roll of 500.
Compliance: DOT 49CFR172.101

Specifications
  • HeaderFLAMMABLE
  • LanguagesEnglish
  • MaterialPaper
  • ComplianceDOT 49 CFR 172.101
  • Width (in)4
  • Height (in)4
  • DescriptionD.O.T. Shipping Labels, Flammable Liquid, 4" x 4"; 500/Roll
Reviews
Q&A