MultiWash 12-Port Coaxial Manifold for III Plate Washers

Representative image only.

MultiWash 12-Port Coaxial Manifold for III Plate Washers

MultiWash | Mfr: 839| Item: 1305428

₹327,000.00INR / Each

₹327,000.00INR / Each
Product Description

Includes

manifold, slip connectors, reservoir kit, two power cords (US standard and European), fuse, rinse strips, dust cover, and, quick-start guide. Order additional manifolds separately.

Key Features

  • Quick-connect fittings allow unit to be fully operational in 15 minutes
  • Rinse-on-demand feature for complete system cleaning at the touch of a button
  • Can handle full range of assay types from the most gentle wash to the most aggressive ELISA

More About this Item

These plate washers feature easy-to-program wash routines, bottom washing, sweeping crosswise aspiration, dispense only and aspirate only functions, and overflow washing for continual dispensing while aspirating off the top for a thorough wash. Use the plate washer to perform cell washing; agglutination tests; cytotoxicity tests; and ELISA, EIA, FIA, and LIA tests.

Minimum residual volume is ≤1 μL per well. Washer has seven programmable wash operations. Independent adjustments of horizontal and vertical needle positions allow any shape well to be accommodated; extra aspiration adjustment for both horizontal and vertical needle positions can be stored in individual programs. Wetted materials include glass, nylon, PTFE, stainless steel, polypropylene, Tygon®, Delrin®, polyethylene, and Santoprene®.

The washer performs a 300-µL three-cycle wash on a full 96-well plate in less than two minutes. You control the dispense pressure. By programming and storing routines, you can switch between seven different modes at the touch of a button. The washer also features a floating manifold that adjusts to match each plate and allows the washer to perform special test procedures. All units are CE-approved, ETL-listed, and certified with CAN/CSA Std C22.2, No. 1010.1.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

ETL ListedTwo year warranty
Specifications
  • Well Volume (mL)2
  • Width (InchShort)16
  • Height (InchShort)11 1/2
  • Number of Channels (Manifold)12
  • Min Residual Volume (mL)<1%/well
  • Max Number of Wash Cycles10
  • Min Dispense Volume (µL)50
  • Accuracy±3% at 300 μL
  • Height (mm)292
  • Description12-Port Coaxial Manifold for III Plate Washers
Reviews
Q&A