MTC Bio Bowtie Biohazard Bags with Drawstring, Autoclavable, 25" x 35", HDPE; 100/CS

Representative image only.

MTC Bio Bowtie Biohazard Bags with Drawstring, Autoclavable, 25" x 35", HDPE; 100/CS

MTC Bio | Mfr: A8001R| Item: 1400303

Drawstring loops makes it easy to close and remove bag from hot autoclave or bag stand

₹50,900.00INR / Case of 100

₹50,900.00INR / Case of 100

Buy It Together

Current Item

MTC Bio Bowtie Biohazard Bags with Drawstring, Autoclavable, 25" x 35", HDPE; 100/CS
MTC Bio Bowtie Biohazard Bags with Drawstring, Autoclavable, 25" x 35", HDPE; 100/CS
1400303

₹50,900.00INR / Case of 100
+ =
Total Price 68400.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Color changing sterilization indicator
  • Clearly marked with a biohazard symbol
  • Large marking area
  • Autoclavable at 250°F (121°C)

More About this Item

Drawstring loops on the autoclavable BowTie biohazard bags offer convenience and safety. Quickly cinch close the bags before autoclaving or disposal. Loops allow for venting during sterilization and a safe way to remove bags from a hot autoclave. A color changing indicator turns dark after sterilization. Bags are made of 2.25 mil thick high-density polyethylene (HDPE) and measure 35"L x 25"W (89 x 64 cm) with a 16 gallon (60 L) capacity.

Order optional BowTie biohazard bin 14003-02 to securely hold the biohazard bags in place for hands-free disposal of hazardous waste.

Specifications
  • Capacity (Gal)16
  • Capacity (Liters)60
  • ColorRed
  • MaterialHigh density polyethylene (HDPE)
  • Length (in)35
  • Width (in)25
  • Length (cm)89
  • Width (cm)64
  • Thickness (mil)2.25
  • DescriptionBowtie™ Biohazard Bags with Drawstring, Autoclavable, 25" x 35", HDPE; 100/CS
Reviews
Q&A