MTC Bio BenchBin™ Benchtop Biohazard Bin

Representative image only.

MTC Bio BenchBin™ Benchtop Biohazard Bin

MTC Bio | Mfr: A8401B| Item: 1400304

Compact footprint—fits on benchtop for easy access

₹8,700.00INR / Each

₹8,700.00INR / Each

Buy It Together

Current Item

MTC Bio BenchBin™ Benchtop Biohazard Bin
MTC Bio BenchBin™ Benchtop Biohazard Bin
1400304

₹8,700.00INR / Each
+
Cole-Parmer Autoclavable Biohazard Bags, Red, 8.5" x 11", PP; 100/PK
1400307

₹10,700.00INR / Pkg of 100
=
Total Price 19400.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Push to open/close lid keeps waste materials covered
  • Use with 8-1/2" x 11" disposal bags
  • Compatible with autoclavable biohazard bags 14003-07

More About this Item

Benchtop biohazard bin only measures 7"L x 5"W x 7"H (18 x 13 x 18 cm) taking up minimal countertop space. Use with 8-1/2" x 11" autoclavable bags 14003-07 for biological waste. Removable cover makes it quick and easy to empty contents.

Specifications
  • Capacity (Liters)1.5
  • Capacity (Gal)0.43
  • MaterialABS plastic/polypropylene
  • ColorRed
  • Compatible WithAutoclavable bags 14003-07
  • Length (InchShort)7
  • Height (in)7
  • Width (InchShort)5
  • Width (cm)13
  • Height (cm)18
  • Length (cm)18
  • DescriptionBenchBin™ Benchtop Biohazard Bin
Reviews
Q&A