Motic Instruments Microscope Camera Adaptor, C-Mount, 0.35X, for 1/3” Chip Sensors
Representative image only.
Motic Instruments Microscope Camera Adaptor, C-Mount, 0.35X, for 1/3” Chip Sensors
Motic Instruments | Mfr: 1101010100861| Item: 4892591
Easily connect your trinocular microscope to a camera
₹43,500.00INR / Each
₹43,500.00INR / Each
Product Description
Key Features
  • Compatible with Motic® trinocular microscopes
  • Compatible with Motic SMZ160-BP, SMZ160-BLED, SMZ160-TP, and SMZ160-TLED trinocular microscopes
More About this Item
In order to use your microscope camera with the trinocular port on your microscope, you need a C-mount adapter to join the microscope and camera together. The C-mount adapter that is needed will depend on your microscope, as well as your camera.
Specifications
  • Compatible WithMotic SMZ160-BP, SMZ160-BLED, SMZ160-TP, and SMZ160-TLED trinocular microscopes
  • ConnectionsC-Mount
  • DescriptionMicroscope Camera Adaptor, C-Mount, 0.35X, for 1/3” Chip Sensors