Motic Instruments Microscope Camera Adaptor, C-Mount, 0.35X, for 1/3” Chip Sensors
Representative image only.
Motic Instruments Microscope Camera Adaptor, C-Mount, 0.35X, for 1/3” Chip Sensors
Motic Instruments | Mfr: 1101001904115| Item: 4892590
Easily connect your trinocular microscope to a camera
₹71,000.00INR / Each
₹71,000.00INR / Each
Product Description
Key Features
  • Compatible with Motic® trinocular microscopes
  • Compatible with Motic BA Series, AE31E/AE2000, and SMZ-161/171 trinocular microscopes
More About this Item
In order to use your microscope camera with the trinocular port on your microscope, you need a C-mount adapter to join the microscope and camera together. The C-mount adapter that is needed will depend on your microscope, as well as your camera.
Specifications
  • Compatible WithMotic BA Series, AE31E/AE2000, and SMZ-161/171 trinocular microscopes
  • ConnectionsC-Mount
  • DescriptionMicroscope Camera Adaptor, C-Mount, 0.35X, for 1/3” Chip Sensors