Morphix 401966-10 Badge Clips for Gas Monitoring Badges; Pk/10
Representative image only.
Morphix 401966-10 Badge Clips for Gas Monitoring Badges; Pk/10
Morphix | Mfr: 401966-10| Item: 8656360
Optimize performance of your ChromAir® Gas Monitoring Badges
₹2,400.00INR / Pkg of 10
₹2,400.00INR / Pkg of 10
Product Description
Key Features
  • After badge is inserted into the color comparator, move the color scale insert until color scale matches color formed on badge
  • Exposed badges give immediate results— no waiting for lab reports
  • Secure gas monitoring badges with these badge clips
  • Never lose a badge again!
More About this Item
Color Comparators increase the range and resolution of your ChromAir badge. The color scales on the comparator identically match the colors formed on the badges.
Badges attach easily to shirt or lab coat collars—order badges separately.
Specifications
  • Compatible WithGas monitoring badges
  • DescriptionBadge clips for Gas Monitoring Badges. Pack of 10