Monarch MI153209 Roll of Reflective Tape for use with Optical Tachometers, 5 ft long and 1/2" wide

Representative image only.

Monarch MI153209 Roll of Reflective Tape for use with Optical Tachometers, 5 ft long and 1/2" wide

Monarch | Mfr: 6180-070| Item: 0821062

Cut and adhere to surface being monitored

₹5,700.00INR / Each

₹5,700.00INR / Each
Product Description

Key Features

  • Ideal for use with Monarch optical tachometers
  • 1 year warranty

More About this Item

All optical tachometers require a reflective surface to operate—use reflective tape to create this surface.

1 Year Warranty
Specifications
  • Length (InchShort)60
  • Width (InchShort)1/2
  • Length (cm)152 2/5
  • Width (cm)1 19/64
  • DescriptionRoll of Reflective Tape for use with Optical Tachometers, 5 ft long and 1/2" wide
  • Warranty1 year
Reviews
Q&A