Monarch 6480-030 Sensor Pk for Examiner 1000
Representative image only.
Monarch 6480-030 Sensor Pk for Examiner 1000
Monarch | Mfr: 6480-030| Item: 9883470
Enhance the versitility of your application with these Monarch Instrument Vibration Accessories
₹235,000.00INR / Each
₹235,000.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Choose from a wide range of accessories for your Monarch Instrument Vibration Meters.
Sensor pack. Includes accelerometer 98834-74, magnetic base 98834-77, connecting stud 98834-81, and stinger 98834-84
1 Year Warranty
Specifications
  • DescriptionVibration Meter Sensor Pk with BNC Connector