Microscope Arm Rests, ESD
Representative image only.
Microscope Arm Rests, ESD
| Mfr: MSM-ESD| Item: 4660002
Enjoy total comfort at your microscope – even after hours of use!
₹70,900.00INR / Pair of 1
₹70,900.00INR / Pair of 1
Product Description
Key Features
  • Soft, padded surface supports forearms, increasing comfort and productivity
  • Use with any size microscope — ideal for most microscope applications
  • 6 month warranty
More About this Item
Put an end to aching muscles and fatigue — these microscope forearm supports reduce static muscle activity, improve working posture, and ensure neutral wrist positioning. The adjustable clamps fit most tables and counters; use with work surfaces from 5/8" to 1-3/4" thick. Adjust the angle of the arm rests to provide the most comfort. Vinyl cover is easy to clean and disinfect.
6 Month Warranty
Specifications
  • DescriptionMicroscope arm rests, ESD