MICROELECTRODE PH BNC
Representative image only.
MICROELECTRODE PH BNC
| Item: 0599300
Take pH measurements in samples as low as 10 µl--even from filter paper and agarose gel
₹216,000.00INR / Each
₹216,000.00INR / Each
Product Description
Key Features
  • Obtain best results in mV or relative mV mode
  • Temperature range is 32 to 167ºF (0 to 75ºC)
  • Immersion depth is 500 µm
  • Compatible with popular pH/mV meters with BNC connectors
  • 6 month warranty
More About this Item
Unbreakable microelectrodes combine measuring and reference cells in a single probe. Tip diameter is less than 1000 µm--fits microwell plates, small capillary tubes, and more. Electrodes are accurate to ±0.01 pH units from 0 to 12 pH. Low impedence provides stable readings. Includes filling solution, plastic dispenser, and manual.
6 Month Warranty
Specifications
  • Electrode TypeSingle junction
  • Electrode Body MaterialUnbreakable Non-Glass Fluoropolymer/PTFE
  • Max Temperature (° C)75
  • ConnectorBNC
  • Diameter (mm)1
  • Length (mm)140
  • DescriptionMicro electrode with BNC
  • Warranty6 month warranty