Mettler Toledo Redox ORP Electrode; S7
Representative image only.
Mettler Toledo Redox ORP Electrode; S7
Mettler Toledo | Mfr: 51343200| Item: 3590230
Precision ORP probes to monitor oxidation-reduction reactions
₹155,000.00INR / Each
₹155,000.00INR / Each
Buy It Together
Current Item
Mettler Toledo Redox ORP Electrode; S7
Mettler Toledo Redox ORP Electrode; S7
3590230
₹155,000.00INR / Each
+ =
Total Price 158300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Cmbination ORP electrodes
  • Cable not included
More About this Item
The ARGENTHAL™ reference system is silver free—the junction won't become blocked by sulfides or proteins.
Select the appropriate sensor cable for your application and meter—BNC, waterproof BNC, DIN, Lemo OO, and wire-only cables available.
Specifications
  • ApplicationsOxidation-reduction
  • Electrode TypePlatinum
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference Electrolyte3M KCl
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • ATC Element
  • ConnectorS7
  • Diameter (mm)12
  • Length (mm)120
  • DescriptionORP Electrode; S7