Mettler Toledo DM405-SC Combined AG2S-Sulfidized Silver Billet Electrode
Representative image only.
Mettler Toledo DM405-SC Combined AG2S-Sulfidized Silver Billet Electrode
Mettler Toledo | Mfr: 51109291| Item: 2575551
Robust electrodes for specialized titration methods and legacy workflows
₹185,000.00INR / Each
₹185,000.00INR / Each
Product Description
Key Features
  • Optimized for specific titration techniques where direct signal response is required
  • High-quality platinum and silver sensing elements deliver reliable, stable measurements
  • Simple, durable construction supports long service life in routine laboratory use
  • Compatible with select Mettler Toledo titrators using classic sensor connections
  • Sulfidized silver billet electrode for sulfide and mercaptan titrations
  • Robust construction for demanding sample matrices
  • Designed for selective argentometric analysis
  • Compatible with Mettler Toledo Excellence titrators and compact potentiometric titrators
More About this Item
These Mettler Toledo classic redox and argentometric electrodes are designed for laboratories that require specialized titration methods or operate legacy titration systems. Their high-quality platinum and silver sensing elements offer direct, uncomplicated signal output for consistent results with method-specific requirements, alternative detection modes, or classic instrument compatibility.
Specifications
  • Electrode MaterialSilver
  • Min temperature (° C)0
  • Max temperature (° C)80
  • Min temperature (° F)32
  • Max temperature (° F)176
  • Use WithExcellence titrators and potentiometric compact titrators
  • DescriptionCombined AG2S-Sulfidized Silver Billet Electrode