Mettler Toledo DGi115-SC Glass pH Electrode w/Ground Glass Junction for Aqueous Media
Representative image only.
Mettler Toledo DGi115-SC Glass pH Electrode w/Ground Glass Junction for Aqueous Media
Mettler Toledo | Mfr: 51109504| Item: 2575557
Versatile glass pH electrodes for precise titration across aqueous and non-aqueous applications
₹271,000.00INR / Each
₹271,000.00INR / Each
Product Description
Key Features
  • Plug & Play sensor chips store calibration and sensor condition data for error-free setup
  • Application-specific membranes and reference systems deliver fast, reproducible results
  • Compatible with Mettler Toledo Excellence and potentiometric compact titrators
  • Optimized for food and beverage quality analysis
  • Fixed ground-glass junction with silver ion trap resists clogging and contamination
  • Fast-response HA glass membrane delivers stable, reproducible measurements
More About this Item
Mettler Toledo DGi Series glass pH electrodes are engineered for reliable potentiometric measurement and acid–base titration across a wide range of laboratory applications. Designs include ceramic pin, ground glass, and movable sleeve junctions to support stable performance in aqueous, low-ionic, and non-aqueous sample matrices.
Integrated Plug & Play sensor chips automatically transfer calibration and sensor status data to compatible titrators, minimizing setup errors and ensuring consistent results. Ideal for demanding pharmaceutical, food, solvent, and micro-volume analyses as well as routine lab measurements.
Specifications
  • ApplicationsAqueous titration; acid/base; pH measurement
  • Electrode TypepH Electrode
  • Electrode Body MaterialGlass
  • Reference JunctionGround glass (fixed)
  • Reference Electrolyte3 mol/L KCl
  • Sensing Glass TypeHA cylindrical
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • OutputPotentiometric
  • ConnectorS12
  • Use WithExcellence titrators; potentiometric compact titrators
  • DescriptionGlass pH Electrode w/Ground Glass Junction for Aqueous Media