Mettler Toledo 30988520 Titration Kit KFC for EVA C1 and C3 Coulometric Karl Fischer Titrators
Representative image only.
Mettler Toledo 30988520 Titration Kit KFC for EVA C1 and C3 Coulometric Karl Fischer Titrators
Mettler Toledo | Mfr: 30988520| Item: 3333226
Simplify Karl Fischer titration methods with titration kits
₹259,000.00INR / Each
₹259,000.00INR / Each
Product Description
Includes
Titration arm, titration arm strap, clear vessel, adapter plate, adapter plate seal, sample injection adapter, septum set (5), NS14 stopper, M9 stopper set (2), 250 g molecular sieves, 1 mL syringe (2), 0.8 x 80 mm injection needle (2), and magnetic stir bar.
Key Features
  • Kits include all essential components for the EVA KF titrators
  • Use with EVA C1 and C3 coulometric titrators
  • 1 year warranty
1 Year Warranty
Specifications
  • Compatible WithEVA C1 and C3 coulometric Karl Fischer titrators
  • DescriptionTitration Kit KFC for EVA C1 and C3 Coulometric Karl Fischer Titrators