Mettler Toledo 30988517 Vessel KFC TS GL80 for EVA Coulometric Karl Fischer Titrators
Representative image only.
Mettler Toledo 30988517 Vessel KFC TS GL80 for EVA Coulometric Karl Fischer Titrators
Mettler Toledo | Mfr: 30988517| Item: 3333227
Enhance your Mettler Toledo EVA volumetric Karl Fischer titrators with specialized vessels
₹309,000.00INR / Each
₹309,000.00INR / Each
Product Description
Key Features
  • Specialized vessels help with dissolution and extraction of samples
  • Use with EVA C1 and C3 coulometric KF titrators
  • Thermostable titration vessel helps improve sample dissolution and extraction
  • Connect to an external oil or water bath to heat sample
  • 1 year warranty
1 Year Warranty
Specifications
  • Compatible WithEVA C1 and C3 coulometric Karl Fischer titrators
  • DescriptionVessel KFC TS GL80 for EVA Coulometric Karl Fischer Titrators, Thermostable