Mettler Toledo 30869302 Vessel KFV L-TS for EVA Volumetric Karl Fischer Titrators
Representative image only.
Mettler Toledo 30869302 Vessel KFV L-TS for EVA Volumetric Karl Fischer Titrators
Mettler Toledo | Mfr: 30869302| Item: 3333217
Enhance your Mettler Toledo EVA volumetric Karl Fischer titrators with specialized vessels
₹330,000.00INR / Each
₹330,000.00INR / Each
Product Description
Key Features
  • Specialized vessels help with dissolution and extraction of samples
  • Use with EVA V1 and V3 volumetric KF titrators
  • Thermostable titration vessel helps improve sample dissolution and extraction
  • Connect to an external oil or water bath to heat sample
  • 1 year warranty
1 Year Warranty
Specifications
  • Compatible WithEVA V1 and V3 volumetric Karl Fischer titrators
  • DescriptionVessel KFV L-TS for EVA Volumetric Karl Fischer Titrators, Large, Thermostatable