Mettler Toledo 30869290 Titration Kit KFV L for EVA V1 and V3 Volumetric Karl Fischer Titrators
Representative image only.
Mettler Toledo 30869290 Titration Kit KFV L for EVA V1 and V3 Volumetric Karl Fischer Titrators
Mettler Toledo | Mfr: 30869290| Item: 3333215
Simplify Karl Fischer titration methods with titration kits
₹282,000.00INR / Each
₹282,000.00INR / Each
Product Description
Includes
Titration arm, titration arm strap, large size vessel, adapter plate, adapter plate seal, sample injection adapter, septum set, M24 stopper, M9 stopper set, drying tube, 250 g molecular sieves, tapered connector, silicone air tube, 5 mL syringe (2), 0.8 x 80 mm injection needle (2), and magnetic stir bar.
Key Features
  • Kits include all essential components for the EVA KF titrators
  • Use with EVA V1 and V3 volumetric titrators
  • 1 year warranty
1 Year Warranty
Specifications
  • Compatible WithEVA V1 and V3 volumetric Karl Fischer titrators
  • DescriptionTitration Kit KFV L for EVA V1 and V3 Volumetric Karl Fischer Titrators