Mettler Toledo 30869288 Burette for Karl Fischer EVA Volumetric Titrators, 10 mL
Representative image only.
Mettler Toledo 30869288 Burette for Karl Fischer EVA Volumetric Titrators, 10 mL
Mettler Toledo | Mfr: 30869288| Item: 3333213
Enhance your Mettler Toledo EVA volumetric Karl Fischer titrators with burettes
₹457,000.00INR / Each
₹457,000.00INR / Each
Product Description
Includes
Burette parking station, drying tube, stopper, dispensing tube, suction tube, flat seal, and tube tip holder.
Key Features
  • Designed specifically for use with EVA V1 and V3 titrators
  • 10 mL capacity
  • 1 year warranty
1 Year Warranty
Specifications
  • Capacity (mL)10
  • Compatible WithEVA V1 and V3 volumetric titrators
  • DescriptionBurette for Karl Fischer EVA Volumetric Titrators, 10 mL