Mettler Toledo 30330848 600 mm Filling Tube for Mettler Densito and DensitoPro

Representative image only.

Mettler Toledo 30330848 600 mm Filling Tube for Mettler Densito and DensitoPro

Mettler Toledo | Mfr: 30330848| Item: 2575333

Enhance the performance of your specific gravity measurement instrument

₹37,900.00INR / Each

₹37,900.00INR / Each
Product Description

Key Features

  • Filling tube, 600 mm L
  • 1 year warranty

More About this Item


Choose from filling tubes, syringes, and more!

1 Year Warranty
Specifications
  • Compatible WithDensity and DensitoPro
  • Length (mm)600
  • Description600 mm Filling Tube for Mettler Densito and DensitoPro
  • Warranty1 year
Reviews
Q&A