Mettler Toledo 12122381 Auxiliary Display For RS232/PL-E Balances
Representative image only.
Mettler Toledo 12122381 Auxiliary Display For RS232/PL-E Balances
Mettler Toledo | Mfr: 12122381| Item: 1114030
Add an extra balance display to create a truly ergonomic workspace
₹190,000.00INR / Each
₹190,000.00INR / Each
Product Description
More About this Item
This auxiliary LCD display with backlighting can be placed up to 10 meters from the balance due to the wireless Bluetooth connection. The extra display can be placed elsewhere on your lab bench, outside a fume cupboard or mounted on a stand or wall.
Specifications
  • DescriptionAuxiliary Display For RS232/PL-E Balances