Mettler Toledo 11107766 Antistatic Kit Power Supply For All Balances
Representative image only.
Mettler Toledo 11107766 Antistatic Kit Power Supply For All Balances
Mettler Toledo | Mfr: 11107766| Item: 1114042
₹767,000.00INR / Each
₹767,000.00INR / Each
Product Description
More About this Item
Static and electrostatic charging can cause inaccuracies, greatly affecting sensitive balances like analytical, micro, and semi-micro versions.
Many tare vessels, especially plastic containers, are prone to electrostatic charging. Powdery samples then scatter or stick to the container from the static electricity. If the sample is toxic or harmful, this could be dangerous to the operator.
Discharge/neutralize your samples before weighing to ensure maximum security of the sample and user, and obtain more accurate measurements.
Specifications
  • DescriptionAntistatic Kit Power Supply For All Balances