Metrepak 270-9.00 pH Calibration Buffer Capsules, 9.0, yellow, 10/vial
Representative image only.
Metrepak 270-9.00 pH Calibration Buffer Capsules, 9.0, yellow, 10/vial
Metrepak | Mfr: 270-9.00| Item: 0597716
No more wasted buffer
₹4,000.00INR / Vial of 10
₹4,000.00INR / Vial of 10
Product Description
Key Features
  • Make only the buffer you need—empty one capsule into 100 mL of water and stir
  • Accurate to ±0.02 pH
  • Ten capsules per vial
More About this Item
Prepare fresh buffer solutions with these handy pH buffer capsules. Empty the contents of one capsule into 100 mL of distilled water and stir. Buffer solutions are accurate to ±0.02 pH. Each vial contains ten capsules.
Specifications
  • Buffer Value9
  • DescriptionpH Calibration Buffer Capsules, 9.0, yellow, 10/vial