Meiji Techno Stereozoom microscope system with boom stand and holder
Representative image only.
Meiji Techno Stereozoom microscope system with boom stand and holder
Meiji Techno | Mfr: EMZ-5/4300+FS/MA502| Item: 4840206
Convenient, preassembled microscope systems provide superb quality and easy ordering
₹709,000.00INR / Each
₹709,000.00INR / Each
Product Description
Key Features
  • System 48402-06 includes:
  • Stereozoom body 48402-20
  • Boom stand and holder 48404-45
  • 10x eyepieces 48402-58
  • Limited Lifetime Warranty
More About this Item
For your added convenience, we also offer preassembled microscope systems that include a microscope body, stand, and eyepieces. All eyepieces have diameters of 34 mm OD x 25 mm ID.
Limited Lifetime Warranty
Specifications
  • Magnification7x to 45x
  • Maximum Magnification45x
  • Head ConfigurationBinocular
  • Objectives0.7x to 4.5x
  • Eyepiece Diameter (mm)34
  • Eyepiece Magnification10x
  • EyepiecesWidefield
  • Stand TypeBoom stand
  • DescriptionStereozoom microscope system with boom stand and holder
  • WarrantyLTD Life Time