Meiji Techno S-4300+FS Boom Microscope Stand; can be tilted 180°
Representative image only.
Meiji Techno S-4300+FS Boom Microscope Stand; can be tilted 180°
Meiji Techno | Mfr: S-4300+FS| Item: 4840445
Custom design your own microscope
₹245,000.00INR / Each
₹245,000.00INR / Each
Product Description
Key Features
  • Choose the microscope stand that best suits your application
  • Limited Lifetime Warranty
More About this Item
Use the components below to build a microscope that best suits your application. We offer a variety of microscope bodies, stands, eyepieces, and ring lights. Components are interchangeable—mix and match to design your perfect microscope! Choose from basic microscope stand, standard microscope stands, or boom microscope stands.
Order microscope and eyepieces separately.
Boom Microscope Stands are ideal when wide horizontal and vertical viewing are important. Stands have their own focusing slide block and illuminator port, and feature a 20-7/8''L horizontal pole and a 15-3/4''L vertical pole. Easily adjust the height on the vertical pole and set it to wherever you need it with the clamp screw. Base measures 10-7/16''L x 10-7/16''W x 1''H. Model 48404-45 can also be titled 180° for added flexibility.
Limited Lifetime Warranty
Specifications
  • Stand TypeBoom stand
  • DescriptionBoom Microscope Stand; can be tilted 180°
  • WarrantyLTD Life Time