Meiji Techno Replacement Microscope Lamp, Halogen, 6V, 30W

Representative image only.

Meiji Techno Replacement Microscope Lamp, Halogen, 6V, 30W

Meiji Techno | Mfr: MA326| Item: 4840151

Optimize life and performance of your Meiji Techno microscopes

₹7,900.00INR / Each

₹7,900.00INR / Each
Product Description

Key Features

  • Easily replacement microscope bulb to keep your microscope working
  • For the 48401-series professional compound microscopes
  • Limited Lifetime Warranty

More About this Item

Keep your Meiji Techno microscopes running smoothly with these replacement bulbs.

Limited Lifetime Warranty
Specifications
  • Illuminator(s)Halogen
  • Power6V, 30W
  • DescriptionReplacement Microscope Lamp, Halogen, 6V, 30W
  • WarrantyLTD Life Time
Reviews
Q&A