Meiji Techno Replacement Microscope Lamp, Halogen, 6V, 10W

Representative image only.

Meiji Techno Replacement Microscope Lamp, Halogen, 6V, 10W

Meiji Techno | Mfr: MA570| Item: 4840250

Optimize life and performance of your Meiji Techno microscopes

₹15,600.00INR / Each

₹15,600.00INR / Each
Product Description

Key Features

  • Easily replacement microscope bulb to keep your microscope working

More About this Item

Keep your Meiji Techno microscopes running smoothly with these replacement bulbs.

For incident and transmitted light of Meiji Techno ABE, ABZH and PBH stands.

Specifications
  • Illuminator(s)Halogen
  • Power6V, 10W
  • DescriptionReplacement Microscope Lamp, Halogen, 6V, 10W
Reviews
Q&A