Meiji Techno Microscopy Immersion Oil, 7.4 mL

Representative image only.

Meiji Techno Microscopy Immersion Oil, 7.4 mL

Meiji Techno | Mfr: MA700| Item: 4840195

Optimize life and performance of your Meiji Techno microscopes

₹5,300.00INR / Each

₹5,300.00INR / Each
Product Description

Key Features

  • Keep your Meiji Techno microscopes running smoothly
  • Designed for use with Oil Immersion Microscope Objectives

More About this Item

Choose from a wide range of accessories and replacement parts to enhance performance and optimize the life of your Meiji Techno microscopes.

Type A immersion oil has a viscosity of 150 cSt and a refractive index of 1.5239. Use with compound microscopes that have a 100x immersion objective.

Lifetime Warranty
Specifications
  • Capacity (mL)7.4
  • Compatible WithOil immersion microscope objectives
  • DescriptionMicroscopy Immersion Oil, 7.4 mL
Documents & Certificates
Reviews
Q&A