Meiji Techno Microscope Camera Adaptor, C-Mount, No Lens
Representative image only.
Meiji Techno Microscope Camera Adaptor, C-Mount, No Lens
Meiji Techno | Mfr: MA151/5N| Item: 4840272
Easily connect your Meiji Techno microscope to a camera
₹47,000.00INR / Each
₹47,000.00INR / Each
Product Description
Key Features
  • Compatible with Meiji Techno microscopes
  • Limited Lifetime Warranty
More About this Item
Use C-mount adapters to attach a digital camera or video camera to the photo tube of your Meiji Techno microscope. The C-mount adapter that is needed will depend on your microscope, as well as your camera. Camera and microscope sold separately.
Limited Lifetime Warranty
Specifications
  • Eyepiece MagnificationNone
  • SizeFits over 25 mm phototube
  • DescriptionMicroscope Camera Adaptor, C-Mount, No Lens
  • WarrantyLTD Life Time