Meiji Techno EMZ-5 Stereozoom microscope bodies; objectives, 0.7x to 4.5x
Representative image only.
Meiji Techno EMZ-5 Stereozoom microscope bodies; objectives, 0.7x to 4.5x
Meiji Techno | Mfr: EMZ-5| Item: 4840220
₹369,000.00INR / Each
₹369,000.00INR / Each
Product Description
Key Features
  • Limited Lifetime Warranty
More About this Item
Get crisp, high-resolution images with excellent stereoscopic viewing. The 45° inclined head has adjustable diopter and interpupillary controls for easy observing. Rotate body a full 360° and lock into any position. The eyepiece diameters on all models are 25 mm ID x 34 mm OD.
Model 48402-20 has convenient side-mounted zoom controls for increased efficiency.
Limited Lifetime Warranty
Specifications
  • Magnification0.7 to 4.5x
  • Maximum Magnification4.5x
  • Head ConfigurationBinocular
  • Objectives0.7x to 4.5x
  • DescriptionStereozoom microscope bodies; objectives, 0.7x to 4.5x
  • WarrantyLTD Life Time