Meiji Techno EMT-1 Dual-power microscope bodies; objectives, 1x/2x
Representative image only.
Meiji Techno EMT-1 Dual-power microscope bodies; objectives, 1x/2x
Meiji Techno | Mfr: EMT-1| Item: 4840320
₹209,000.00INR / Each
₹209,000.00INR / Each
Product Description
Key Features
  • Limited Lifetime Warranty
More About this Item
Get crisp, high-resolution images with excellent stereoscopic viewing. The 45° inclined head has adjustable diopter and interpupillary controls for easy observing. Rotate body a full 360° and lock into any position. The eyepiece diameters on all models are 25 mm ID x 34 mm OD.
Dual-power models feature paired objectives mounted in a revolving turret for easy switching between low and high magnification.
Limited Lifetime Warranty
Specifications
  • Magnification1x/2x
  • Maximum Magnification2x
  • Head ConfigurationBinocular
  • Objectives1x/2x
  • DescriptionDual-power microscope bodies; objectives, 1x/2x
  • WarrantyLTD Life Time