Medium Bottle/Cylinder Brush, 16"; 12/PK

Representative image only.

Medium Bottle/Cylinder Brush, 16"; 12/PK

| Item: 8455016

₹25,100.00INR / Pkg of 12

₹25,100.00INR / Pkg of 12

Buy It Together

Current Item

Medium Bottle/Cylinder Brush, 16"; 12/PK
Medium Bottle/Cylinder Brush, 16"; 12/PK
8455016

₹25,100.00INR / Pkg of 12
+ =
Total Price 30900.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 1 month warranty

More About this Item

Ideal for cleaning bottles, cylinders, and large test tubes. Full radial end tip promotes effective cleaning and reduces scratching of glass and plasticware. Wire handle has hanging hook.

1 Month Warranty
Specifications
  • MaterialNylon
  • ColorBlack
  • Handle Length (in)11
  • DescriptionMedium Bottle/cylinder Brush, 16", 12/pk
  • Warranty1 month
Reviews
Q&A