Master Appliance MT-76K Trigger Torch Heat Kit
Representative image only.
Master Appliance MT-76K Trigger Torch Heat Kit
Master Appliance | Mfr: MT-76K| Item: 0303053
Hand-held or table-top butane torch
Call For Price
Call For Price
Product Description
Key Features
  • Meets all consumer products safety commission (CPSC) requirements
  • Adjustable broad or pinpoint flame
More About this Item
This refillable microtorch produces an adjustable wind resistant flame with temperatures up to 2500°F (1300°C). The pinpoint flame is perfect for heat shrinking and terminating as well as general heating applications. The torch features a built-in self-igniting system with gas shut-off safety lock, a extra wide base perfect for hands-free use and a durable fuel tank containing enough fuel for up to 2 hours of usage. Torch measure 6.1" H x 3" W x 1.3" D
Specifications
  • Gas TypeButane
  • Width (in)1 1/4
  • Length (in)3 3/4
  • Height (in)6 1/2
  • DescriptionTrigger Torch Heat Kit
  • WarrantyManufacture Defect