Mark-10 G1023 Bottle Grip for Series TST Stands, 0.050” - 6.000” dia.
Representative image only.
Mark-10 G1023 Bottle Grip for Series TST Stands, 0.050” - 6.000” dia.
Mark-10 | Mfr: G1023| Item: 5984835
Securely grips bottles and containers for closure torque testing
₹168,000.00INR / Each
₹168,000.00INR / Each
Product Description
Key Features
  • Torque capacity of 100 lbf in
  • Ideal for use with TST AND TSTM-DC
  • 3 year warranty
More About this Item
These grips are a key component of testing systems, designed for a wide range of torque applications.
This bottle grip features eight rubber edged gripping arms to secure a wide range of sample shapes and sizes for closure torque testing. The grips can be individually positioned at angles up to 180°. Mounts directly to Series TST torque test stands, most force test stands, and G1045 90° peel fixture.
3 Year Warranty
Specifications
  • DescriptionBottle grip for 59848-00 and -02 test stands (0.050"-6.00" diameter)