Mark-10 AF022 Profiles
Representative image only.
Mark-10 AF022 Profiles
Mark-10 | Mfr: AF022| Item: 2530332
Configurable for your application needs!
₹179,000.00INR / Each
₹179,000.00INR / Each
Product Description
Key Features
  • Requires Mark-10 Motorized Test Stand Before Purchase
More About this Item
Save and recall sets of test parameters, such as speeds, travel limits, preloads, etc. Up to 50 profiles may be stored.
Requires:
- Nothing additional
Specifications
  • DescriptionPush / Pull Test Stand Add-on Option; Profiles