Mark-10 AF021 Preload / Sample Touch
Representative image only.
Mark-10 AF021 Preload / Sample Touch
Mark-10 | Mfr: AF021| Item: 2530331
Configurable for your application needs!
₹89,300.00INR / Each
₹89,300.00INR / Each
Product Description
Key Features
  • Requires Mark-10 Motorized Test Stand Before Purchase
More About this Item
Stops the crosshead and/or zeroes the travel display at an initial preload - useful in tensile, compression, spring, elongation, and other applications. Preload is programmable as a percentage of force gauge/load cell full scale. Three modes: (1) stop, (2) stop and zero, and (3) zero without stopping.
Requires:
- 25303-20 (AF009) travel indication option
- Series 7 or 5 force gauge with load cell
- 25303-16 (09-1214) interface cable
Specifications
  • DescriptionPush / Pull Test Stand Add-on Option; Preload