Mark-10 AF020 Load Holding
Representative image only.
Mark-10 AF020 Load Holding
Mark-10 | Mfr: AF020| Item: 2530330
Configurable for your application needs!
₹140,000.00INR / Each
₹140,000.00INR / Each
Product Description
Key Features
  • Requires Mark-10 Motorized Test Stand Before Purchase
More About this Item
Dynamically adjusts the crosshead position to maintain a programmed load for an indefinite or specified period of time.
Requires:
- Series 7 or 5 force gauge with load cell
If a specified time is required:
- 25303-25 (AF014) cycling / dwell time option
- 25303-16 (09-1214) interface cable
Specifications
  • DescriptionPush / Pull Test Stand Add-on Option; Load Holding