Mark-10 AF014 Cycling / Dwell Time
Representative image only.
Mark-10 AF014 Cycling / Dwell Time
Mark-10 | Mfr: AF014| Item: 2530325
Configurable for your application needs!
₹89,300.00INR / Each
₹89,300.00INR / Each
Product Description
Key Features
  • Requires Mark-10 Motorized Test Stand Before Purchase
More About this Item
The crosshead moves to a limit switch, force set point, travel position, or break, then stops and reverses direction at full speed to the opposite limit. And, the ability to program up to 100,000 cycles. Programmable dwell time for upper and lower limits (set independently) can be set up for up to 10,000 seconds.
Requires:
- If reversing at force set point: Series 7 or 5 force gauge with load cell, and 25303-16 (09-1214) interface cable
- If reversing at travel limit: 25303-22 (AF011) programmable travel limits option
- If reversing at break: 25303-29 (AF019) break detection option and 25303-16 (09-1214) interface cable
- If independent up and down speeds are wanted: 25303-26 (AF016) independent up and down speed option
Specifications
  • DescriptionPush / Pull Test Stand Add-on Option; Cycle/Dwell Time