Mark-10 AF013 Auto Return
Representative image only.
Mark-10 AF013 Auto Return
Mark-10 | Mfr: AF013| Item: 2530324
Configurable for your application needs!
₹62,300.00INR / Each
₹62,300.00INR / Each
Product Description
Key Features
  • Requires Mark-10 Motorized Test Stand Before Purchase
More About this Item
The crosshead moves to a limit switch, force set point, travel position, or break, then stops and reverses direction at full speed to the opposite limit.
Requires:
- If reversing at force set point: Series 7 or 5 force gauge with load cell, and 25303-16 (09-1214) interface cable
- If reversing at travel limit: 25303-22 (AF011) programmable travel limits option
- If reversing at break: 25303-29 (AF019) break detection option and 25303-16 (09-1214) interface cable
Specifications
  • DescriptionPush / Pull Test Stand Add-on Option; Auto Return