Mark-10 AF012 Integrated Overload Protection
Representative image only.
Mark-10 AF012 Integrated Overload Protection
Mark-10 | Mfr: AF012| Item: 2530323
Configurable for your application needs!
₹89,300.00INR / Each
₹89,300.00INR / Each
Product Description
Key Features
  • Requires Mark-10 Motorized Test Stand Before Purchase
More About this Item
Protects a force gauge or force sensor against overload. Program the desired percentage of full scale of the gauge. Adjustable analog voltage setting allows the stand to interface with virtually any gauge with analog output.
Requires:
- Series 7 or 5 force gauge with load cell
- 25303-17 (09-1214) interface cable
Specifications
  • DescriptionPush / Pull Test Stand Add-on Option; Overload Protection