Mark-10 AF009 Travel Indication with USB Output
Representative image only.
Mark-10 AF009 Travel Indication with USB Output
Mark-10 | Mfr: AF009| Item: 2530320
Configurable for your application needs!
₹326,000.00INR / Each
₹326,000.00INR / Each
Product Description
Key Features
  • Requires Mark-10 Motorized Test Stand Before Purchase
More About this Item
Travel is indicated on the control unit display, with output via USB. An internal scale utilizing Renishaw technology produces significantly higher accuracy than with conventional rotary encoderbased designs. Backlash and nonlinearity are virtually eliminated.
Requires:
- Field hardware installation required (if not ordered at the same time as test stand).
- Series 7 or 5 force gauge with load cell.
If data output to a PC is also wanted:
-25303-16 (09-1214) and 25303-17 (09-1158) cables
Specifications
  • DescriptionPush / Pull Test Stand Add-on Option; Travel Measurement