MAPA Butoflex 650 Cotton-Lined Butyl Gloves, size 8, 1 pair
Representative image only.
MAPA Butoflex 650 Cotton-Lined Butyl Gloves, size 8, 1 pair
MAPA | Mfr: 65031-8| Item: 8635741
Inner fabric lining provides exceptional comfort
₹24,200.00INR / Pair of 1
₹24,200.00INR / Pair of 1
Product Description
More About this Item
Butyl gloves provide excellent chemical resistance in applications involving acids, ketones, esters, and amine derivatives. 100% seamless cotton liner provides extra comfort for long-term use. Scalloped cuffs can be rolled down to prevent drips from reaching your arms. Unique Z-pattern grip design for optimal grip.
Specifications
  • MaterialButyl
  • Thickness (mil)60
  • Size8
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • LiningCotton
  • CuffScalloped
  • SterileNo
  • DescriptionCotton-Lined Butyl Gloves, size 8, 1 pair