MAPA 517317 Cleanroom Z-Grip Gloves, size 7-7.5, 12 pairs/pack
Representative image only.
MAPA 517317 Cleanroom Z-Grip Gloves, size 7-7.5, 12 pairs/pack
MAPA | Mfr: 517317| Item: 4021401
Added thickness for mixing, handling, and moving chemicals
₹9,800.00INR / Pkg of 12
₹9,800.00INR / Pkg of 12
Product Description
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Gloves feature a "Z" pattern to enhance grip when handling wet items. Made of natural rubber, nitrile, and neoprene. Individually packaged per pair.
Compliance: ISO Class 4
Conformite Europeenne
Specifications
  • MaterialNitrile
  • Thickness (mil)20
  • Size7 to 7.5
  • Length (in)14
  • Length (cm)35
  • ColorCream
  • GripZ-pattern
  • ComplianceISO Class 4
  • SterileNo
  • DescriptionCleanroom Z-Grip Gloves, size 7-7.5, 12 pairs/pack