MAPA 332429 Heat-Insulated Neoprene Gloves, Large
Representative image only.
MAPA 332429 Heat-Insulated Neoprene Gloves, Large
MAPA | Mfr: 332429| Item: 4021131
Two-tone wear indicator warns you when to replace gloves
₹9,500.00INR / Pair of 1
₹9,500.00INR / Pair of 1
Product Description
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
These gloves are made of synthetic neoprene rubber for good chemical resistance. Easily maintain your grip with the nonslip, roughened surface. Withstand temperatures from -100 to 480°F (-73 to 249°C). The 14"L gloves feature integral cotton fleece liner.
Conformite Europeenne
Specifications
  • MaterialNeoprene
  • SizeLarge
  • Length (in)14
  • Length (cm)35
  • ColorBlack and Blue
  • LiningCotton
  • ComplianceFDA 21 CFR 177.2600
  • SterileNo
  • DescriptionHeat-Insulated Neoprene Gloves, Large