MAGic Clamp H1000-MR-MP Microplate Magnetic Clamp, Holds 1.
Representative image only.
MAGic Clamp H1000-MR-MP Microplate Magnetic Clamp, Holds 1.
MAGic Clamp | Mfr: H1000-MR-MP| Item: 5170527
Optimize performance of your shaking incubator
₹24,000.00INR / Each
₹24,000.00INR / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
Change the capacitiy and capabilities of your shaker with these parts and accessories. Compatible with Benchmark Incu-Shaker™ Mini Incubating Shakers sold separately as 51705-01 and 51705-03.
The convenient MAGic Clamp™ platform is suitable for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.
Two year warranty
Specifications
  • Capacity (mL)1 Microplate
  • Compatible WithBenchmark Scientific Incu-Shaker™ Mini Incubating Shaker
  • ConnectionsMAGic Clamp™
  • DescriptionMicroplate Magnetic Clamp